tert-Butyl-6-bromopyridin-2-ylcarbamate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (6-bromopyridin-2-yl)carbamate, N-Boc-2-Amino-6-bromopyridine

A265320-1G A265320-5G A265320-100MG A265320-10G A265320-250MG
103812-726EA 25 USD
103812-726 103852-620 77092-302 77092-304 77140-836 77140-838 77140-840
tert-Butyl-6-bromopyridin-2-ylcarbamate 97%
tert-Butyl-6-bromopyridin-2-ylcarbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrN₂O₂
MW: 273.14 g/mol
Boiling Pt: 299.3 °C (760 mm Hg)
Flash Pt: 134.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07367929
CAS Number: 344331-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR