N-Boc-2-amino-4-formylthiazole 97%

Supplier: Ambeed
Warning

Synonyms: N-Boc-2-Amino-4-formylthiazole

A209496-1G A209496-250MG A209496-5G
103817-944EA 52 USD
103817-944 77086-976 77139-572 77139-574
N-Boc-2-amino-4-formylthiazole 97%
N-Boc-2-amino-4-formylthiazole

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₃S
MW: 228.2682 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08275704
CAS Number: 494769-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR