N-Boc-2-(4-aminophenyl)ethanol 97%

Supplier: Ambeed
Warning

Synonyms: N-Boc-2-(4-Aminophenyl)ethanol

A419689-25G A419689-10G A419689-250MG
103827-370EA 18 USD
103827-370 103838-418 76842-034 77033-854 77033-856
N-Boc-2-(4-aminophenyl)ethanol 97%
N-Boc-2-(4-aminophenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO₃
MW: 237.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04974330
CAS Number: 104060-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR