N-(tert-Butoxycarbonyl)-1H-pyrazole-1-carboxamidine 98%

Supplier: Ambeed
Warning

Synonyms: 1-(N-Boc-amidino)pyrazole, N-Boc-1H-pyrazole-1-carboximidamide, 1-[N-(tert-Butoxycarbonyl)carbamimidoyl]pyrazole, N-Boc-1H-pyrazole-1-carboxamidine, N-(tert-Butoxycarbonyl)-1H-pyrazole-1-carboximidamide, 1-[N-(tert-Butoxycarbonyl)amidino]pyrazole, N-Boc-1-Guanylpyrazole

A112170-100G A112170-25G A112170-1G A112170-5G
103797-970EA 5 USD
103797-970 103823-806 76718-134 76718-136 77284-216 77310-616
N-(tert-Butoxycarbonyl)-1H-pyrazole-1-carboxamidine 98%
N-(tert-Butoxycarbonyl)-1H-pyrazole-1-carboxamidine

Precaución: for Research Use Only

Formula: C₉H₁₄N₄O₂
MW: 210.24 g/mol
Boiling Pt: 326.2 °C (760 mm Hg)
Melting Pt: 97…101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00216663
CAS Number: 152120-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR