N-Boc-1,6-diaminohexane 97%

Supplier: Ambeed
Warning

Synonyms: N-tert-Butoxycarbonyl-1,6-hexanediamine, N-Boc-1,6-Diaminohexane

A164342-100G A164342-10G A164342-500G A164342-1G A164342-25G A164342-5G
103805-852EA 6 USD
103805-852 103844-112 76799-148 76799-150 76799-152 77320-272 77320-274 77320-276
N-Boc-1,6-diaminohexane 97%
N-Boc-1,6-diaminohexane

Precaución: for Research Use Only

Formula: C₁₁H₂₄N₂O₂
MW: 216.33 g/mol
Flash Pt: 125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00671489
CAS Number: 51857-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR