N-Benzylhydroxylamine hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A103633-1000G
A103633-100G
A103633-10G
A103633-25G
A103633-500G
A103633-5G
103822-294EA
5
USD
103822-294
76717-678
76717-680
76717-682
76717-684
76717-686
77283-136
N-Benzylhydroxylamine hydrochloride 98%
N-Benzylhydroxylamine hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₁₀ClNO Boiling Pt: 253.9 °C (760 mm Hg) Melting Pt: 108…110 °C Flash Pt: 135.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043377 CAS Number: 29601-98-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...