N-Benzyl-2-chloro-N-(2-chloroethyl)ethanamine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Benzyl[bis(2-chloroethyl)]amine hydrochloride

A421232-100G A421232-10G A421232-1G A421232-5G A421232-25G
103827-402EA 5 USD
103827-402 103838-448 76725-252 76725-254 76842-064 76842-066 77307-214
N-Benzyl-2-chloro-N-(2-chloroethyl)ethanamine hydrochloride 95%
N-Benzyl-2-chloro-N-(2-chloroethyl)ethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₆Cl₃N
MW: 268.62 g/mol
Melting Pt: 147…150 °C
Flash Pt: 100.5 °C
Storage Temperature: Freezer
MDL Number: MFCD00185654
CAS Number: 10429-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR