N-Benzoyl-(2R,3S)-3-Phenylisoserine 97%

Supplier: Ambeed
Warning

Synonyms: (2R,3S)-3-Benzamido-2-Hydroxy-3-Phenylpropanoic Acid

A100375-10G A100375-250MG A100375-1G A100375-5G A100375-25G
103795-406EA 67 USD
103795-406 103821-104 77107-156 77107-158 77296-658 77296-660 77684-924
N-Benzoyl-(2R,3S)-3-Phenylisoserine 97%
N-Benzoyl-(2R,3S)-3-Phenylisoserine

Precaución: for Research Use Only

Formula: C₁₆H₁₅NO₄
MW: 285.29 g/mol
Boiling Pt: 580.7 °C (760 mm Hg)
Melting Pt: 169…172 °C
Flash Pt: 305 °C
Storage Temperature: Ambient
MDL Number: MFCD00274633
CAS Number: 132201-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR