1,8-Dihydroxynaphthalene 98%

Supplier: Ambeed
Danger

A278975-5G A278975-10G
103813-442EA 41 USD
103813-442 103853-386 77568-554 77593-718
1,8-Dihydroxynaphthalene 98%
1,8-Dihydroxynaphthalene

Precaución: For research use only. Inert atmosphere, room temperature

Formula: C₁₀H₈O₂
Melting Pt: 137…143 °C
Flash Pt: 193.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00042701
CAS Number: 569-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR