1,5-Dihydroxynaphthalene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,5-Naphthalenediol, Naphthalene-1,5-diol
A226470-25G
A226470-100G
A226470-500G
103809-602EA
75
USD
103809-602
77088-884
77193-012
77193-014
1,5-Dihydroxynaphthalene 98%
1,5-Dihydroxynaphthalene
Precaución: for Research Use Only
Formula:
C₁₀H₈O₂ MW: 160.17 g/mol Boiling Pt: 375.4 °C (760 mm Hg) Melting Pt: 259…261 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003980 CAS Number: 83-56-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...