1,3-Dihydroxynaphthalene 98%

Supplier: Ambeed
Warning

A194834-100G A194834-100MG A194834-10G A194834-250MG A194834-1G A194834-25G A194834-5G
103816-696EA 10 USD
103816-696 103847-310 76800-138 76800-140 76800-142 76800-144 77085-240 77085-242 77085-244
1,3-Dihydroxynaphthalene 98%
1,3-Dihydroxynaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₈O₂
Melting Pt: 123…125 °C
Flash Pt: 185.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00003965
CAS Number: 132-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR