Naphthalen-2-ylhydrazine hydrochloride 95%

Supplier: Ambeed
Warning

A221089-100G A221089-10G A221089-500G A221089-25G A221089-5G
103849-772EA 6 USD
103849-772 76999-914 76999-916 76999-918 77088-368 77303-578
Naphthalen-2-ylhydrazine hydrochloride 95%
Naphthalen-2-ylhydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClN₂
Boiling Pt: 362.9 °C (760 mm Hg)
Melting Pt: 123 °C
Flash Pt: 203.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04109481
CAS Number: 2243-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR