Naphthalen-2-yl diphenyl phosphate 97%

Supplier: Ambeed
Warning

A180470-100G A180470-10G A180470-250MG A180470-1G A180470-5G
103807-540EA 12 USD
103807-540 103845-896 76824-866 76824-868 76824-870 77083-390 77083-392
Naphthalen-2-yl diphenyl phosphate 97%
Naphthalen-2-yl diphenyl phosphate

Precaución: for Research Use Only

Formula: C₂₂H₁₇O₄P
MW: 376.34 g/mol
Boiling Pt: 469.3 °C (760 mm Hg)
Flash Pt: 250.9 °C
Storage Temperature: Ambient
MDL Number: MFCD22380593
CAS Number: 18872-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR