1-Naphthalenemethanol 98%

Supplier: Ambeed
Warning

Synonyms: Naphthalen-1-ylmethanol

A179694-25G A179694-5G A179694-100G A179694-10G
103845-808EA 8 USD
103845-808 77083-278 77083-280 77236-518 77236-520
1-Naphthalenemethanol 98%
1-Naphthalenemethanol

Precaución: for Research Use Only

Formula: C₁₁H₁₀O
MW: 158.20 g/mol
Boiling Pt: 303.6 °C (760 mm Hg)
Melting Pt: 61…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00004044
CAS Number: 4780-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR