1-Naphthylmethylamine 98%

Supplier: Ambeed
Warning

Synonyms: 1-Naphthalenemethylamine, Naphthalen-1-ylmethanamine, Naphthalene-1-methylamine, 1-(Aminomethyl)naphthalene

A437499-1G A437499-5G A437499-10G A437499-100G
103827-626EA 11 USD
103827-626 103838-692 76842-430 76842-432 77572-356 77879-961
1-Naphthylmethylamine 98%
1-Naphthylmethylamine

Precaución: for Research Use Only

Formula: C₁₁H₁₁N
Boiling Pt: 290…293 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004048
CAS Number: 118-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR