1-Naphthyl boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1-Naphthaleneboronic acid, Naphthalene-1-boronic acid, Naphthalen-1-ylboronic acid

A280815-100G A280815-10G A280815-1G A280815-500G A280815-25G A280815-5G
103853-474EA 6 USD
103853-474 76802-172 76802-174 76802-176 76802-178 76837-166 77305-366
1-Naphthyl boronic acid 98%
1-Naphthyl boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₉BO₂
Melting Pt: 208…214 °C
Flash Pt: 184.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00019722
CAS Number: 13922-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR