NAD+ 98%

Supplier: Ambeed
Warning

Synonyms: 1-((2R,3R,4S,5R)-5-((((((((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)oxidophosphoryl)oxy)methyl)-3,4-dihydroxytetrahydrofuran-2-yl)-3-carbamoylpyridin-1-ium

A141009-10G A141009-250MG A141009-1G A141009-25G A141009-5G A141009-100G
103802-932EA 19 USD
103802-932 76798-260 76798-262 77316-060 77316-062 77316-064 77600-048
NAD+ 98%
NAD+

Precaución: for Research Use Only

Formula: C₂₁H₂₇N₇O₁₄P₂
MW: 663.425 g/mol
Melting Pt: 140…142 °C
Storage Temperature: Freezer
MDL Number: MFCD00036253
CAS Number: 53-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR