N-Acetylneuraminic acid 97%

Supplier: Ambeed
Warning

Synonyms: Neu5Ac, (4S,5R,6R,7S,8R)-5-Acetamido-4,6,7,8,9-pentahydroxy-2-oxononanoic acid, NANA, Sialic acid, 5-Acetamido-3,5-dideoxy-D-glycero-D-galacto-nonulopyranosonic acid, Lactaminic acid

A119785-25G A119785-1G A119785-5G A119785-100G A119785-10G
103799-836EA 9 USD
103799-836 76797-282 77311-936 77311-938 77566-216 77566-218
N-Acetylneuraminic acid 97%
N-Acetylneuraminic acid

Precaución: For research use only.

Formula: C₁₁H₁₉NO₉
Boiling Pt: 395.6 °C (760 mm Hg)
Melting Pt: 184…186 °C
Storage Temperature: Freezer
MDL Number: MFCD00006620
CAS Number: 131-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR