N-Acetylcytidine 97%

Supplier: Ambeed
Warning

Synonyms: N-Acetyl-1-(β-D-ribofuranosyl)cytosine, N-{1,2-Dihydro-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl]-2-oxopyrimidin-4-yl}acetamide, N-Ethanoylcytidine, N-[1,2-Dihydro-2-oxo-1-(β-D-ribofuranosyl)pyrimidin-4-yl]acetamide, N4-Acetylcytidine

A270613-1G A270613-5G A270613-10G A270613-100G
103813-008EA 6 USD
103813-008 103852-914 77092-678 77092-680 77276-768 77550-684
N-Acetylcytidine 97%
N-Acetylcytidine

Precaución: for Research Use Only

Formula: C₁₁H₁₅N₃O₆
MW: 285.253 g/mol
Melting Pt: 199 °C
Storage Temperature: Freezer
MDL Number: MFCD00006540
CAS Number: 3768-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR