N6-Benzoyladenosine 95%

Supplier: Ambeed
Warning

A246098-100G A246098-10G A246098-1G A246098-25G A246098-5G A246098-500G
103811-686EA 5 USD
103811-686 103851-516 77031-366 77031-368 77090-794 77090-796 77090-798 77600-982
N6-Benzoyladenosine 95%
N6-Benzoyladenosine

Precaución: for Research Use Only

Formula: C₁₇H₁₇N₅O₅
MW: 371.35 g/mol
Melting Pt: 141…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00056988
CAS Number: 4546-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR