N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxyadenosine 95%

Supplier: Ambeed
Warning

Synonyms: Bz-DMT-dA, N-(9-((2R,4S,5R)-5-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-hydroxytetrahydrofuran-2-yl)-9H-purin-6-yl)benzamide

A302183-10G A302183-250MG A302183-1G A302183-25G A302183-5G A302183-100G
103814-330EA 16 USD
103814-330 103835-486 76816-790 76816-792 76838-174 77305-846 77305-848 77612-738
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxyadenosine 95%
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxyadenosine

Precaución: for Research Use Only

Formula: C₃₈H₃₅N₅O₆
MW: 657.73 g/mol
Melting Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00010058
CAS Number: 64325-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR