N6-(3-Methyl-2-butenyl)adenosine 98%

Supplier: Ambeed
Warning

Synonyms: N6-Isopentenyladenosine, (2R,3S,4R,5R)-2-(Hydroxymethyl)-5-(6-((3-methylbut-2-en-1-yl)amino)-9H-purin-9-yl)tetrahydrofuran-3,4-diol

A274098-250MG A274098-1G A274098-100MG
103799-550EA 39 USD
103799-550 103813-182 103853-106 76723-114 77092-926 77601-212
N6-(3-Methyl-2-butenyl)adenosine 98%
N6-(3-Methyl-2-butenyl)adenosine

Precaución: for Research Use Only

Formula: C₁₅H₂₁N₅O₄
MW: 335.35 g/mol
Boiling Pt: 647.2±65.0 °C (760 mm Hg)
Melting Pt: 143…146 °C
Flash Pt: 345.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005741
CAS Number: 7724-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR