N4-(3-Chloro-4-fluorophenyl)-7-methoxyquinazoline-4,6-diamine 97%

Supplier: Ambeed
Warning

A200880-5G A200880-250MG A200880-25G A200880-1G
103817-250EA 69 USD
103817-250 103847-884 77085-950 77237-138 77237-140 77302-728
N4-(3-Chloro-4-fluorophenyl)-7-methoxyquinazoline-4,6-diamine 97%
N4-(3-Chloro-4-fluorophenyl)-7-methoxyquinazoline-4,6-diamine

Precaución: for Research Use Only

Formula: C₁₅H₁₂ClFN₄O
MW: 318.73 g/mol
Boiling Pt: 487.3±45.0 °C (760 mm Hg)
Flash Pt: 248.5 °C
Storage Temperature: Ambient
MDL Number: MFCD30342451
CAS Number: 179552-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR