N2-Methylpyridine-2,5-diamine 98%
Supplier: Ambeed
|
Warning
|
A192412-100MG
A192412-250MG
103816-438EA
281
USD
103816-438
76721-160
76721-162
N2-Methylpyridine-2,5-diamine 98%
Precaución: for Research Use Only
Formula:
C₆H₉N₃ Boiling Pt: 306.4 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD08692343 CAS Number: 28020-36-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...