N2-Methyl-2,3-pyridinediamine 97%
Supplier: Ambeed
|
Warning
|
A109210-100MG
A109210-250MG
A109210-5G
103797-466EA
62
USD
103797-466
76970-620
76970-622
76970-624
N2-Methyl-2,3-pyridinediamine 97%
Precaución: for Research Use Only
Formula:
C₆H₉N₃ Boiling Pt: 303.553 °C (760 mm Hg) Flash Pt: 137.384 °C Storage Temperature: Ambient |
MDL Number:
MFCD08692132 CAS Number: 5028-20-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...