N2-Isobutyryl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxyguanosine 95%

Supplier: Ambeed
Warning

Synonyms: 5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-deoxyguanosine, iBu-DMT-dG, N2-iBu-DMT-dG

A135681-10G A135681-5G A135681-1G A135681-25G A135681-100G
103802-204EA 7 USD
103802-204 76811-490 76811-492 77298-810 77315-050 77315-052 77883-097
N2-Isobutyryl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxyguanosine 95%
N2-Isobutyryl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxyguanosine

Precaución: for Research Use Only

Formula: C₃₅H₃₇N₅O₇
MW: 639.71 g/mol
Melting Pt: 150 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010059
CAS Number: 68892-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR