N-Phenyl-o-phenylenediamine 98%

Supplier: Ambeed
Warning

Synonyms: N-(2-Aminophenyl)aniline, 2-Aminodiphenylamine, N1-Phenylbenzene-1,2-diamine

A202099-100G A202099-10G A202099-500G A202099-25G A202099-5G
103847-990EA 7 USD
103847-990 76770-180 76770-182 76770-184 77086-086 77302-790
N-Phenyl-o-phenylenediamine 98%
N-Phenyl-o-phenylenediamine

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂
Melting Pt: 75 °C
Flash Pt: 181.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00007685
CAS Number: 534-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR