N1-Methyl-N1-phenylethane-1,2-diamine dihydrochloride 95%

Supplier: Ambeed
Warning

A295525-100MG A295525-250MG A295525-1G A295525-5G
103814-062EA 135 USD
103814-062 103835-196 76723-558 76723-560 77305-672 77305-674
N1-Methyl-N1-phenylethane-1,2-diamine dihydrochloride 95%
N1-Methyl-N1-phenylethane-1,2-diamine dihydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₆Cl₂N₂
MW: 223.14 g/mol
Boiling Pt: 247.5 °C (760 mm Hg)
Melting Pt: 205 °C
Storage Temperature: Ambient
MDL Number: MFCD07841700
CAS Number: 36271-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR