N-Methyl-1,2-phenylenediamine 97%

Supplier: Ambeed
Warning

Synonyms: N1-Methylbenzene-1,2-diamine, N-Methylbenzene-1,2-diamine, N-Methyl-o-phenylenediamine, 2-Amino-N-methylaniline

A109983-100G A109983-10G A109983-25G A109983-1G A109983-5G
103797-600EA 9 USD
103797-600 103823-408 76970-674 76970-676 76970-678 77283-914 77310-216
N-Methyl-1,2-phenylenediamine 97%
N-Methyl-1,2-phenylenediamine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MW: 122.171 g/mol
Boiling Pt: 252.4±23.0 °C (760 mm Hg)
Melting Pt: 22 °C
Flash Pt: 122.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00192314
CAS Number: 4760-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR