N1-Methyl-4-nitro-1,2-phenylenediamine 98%

Supplier: Ambeed
Warning

Synonyms: 1,2-Diamino-N1-methyl-4-nitrobenzene, N1-Methyl-4-nitrobenzene-1,2-diamine, 2-Amino-1-methylamino-4-nitrobenzene

A140374-5G A140374-1G
103802-862EA 8 USD
103802-862 77299-134 77315-978
N1-Methyl-4-nitro-1,2-phenylenediamine 98%
N1-Methyl-4-nitro-1,2-phenylenediamine

Precaución: for Research Use Only

Formula: C₇H₉N₃O₂
Boiling Pt: 366.8 °C (760 mm Hg)
Melting Pt: 174…176 °C
Flash Pt: 175.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00156607
CAS Number: 41939-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR