N,N'-Dimethyl-1,4-butanediamine 95%

Supplier: Ambeed
Danger

Synonyms: N1,N4-Dimethylbutane-1,4-diamine

A205652-100MG A205652-250MG A205652-1G A205652-100G
103817-652EA 20 USD
103817-652 103848-318 76721-550 76721-552 77086-542 77667-720
N,N'-Dimethyl-1,4-butanediamine 95%
N,N'-Dimethyl-1,4-butanediamine

Precaución: for Research Use Only

Formula: C₆H₁₆N₂
MW: 116.205 g/mol
Boiling Pt: 158.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00039799
CAS Number: 16011-97-5
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR