N,N',N''-Trimethyldiethylenetriamine 97%

Supplier: Ambeed
Warning

Synonyms: 2,2'-Bis(methylamino)-N-methyldiethylamine, N,N-Bis[2-(methylamino)ethyl]methylamine, N1,N2-Dimethyl-N1-(2-(methylamino)ethyl)ethane-1,2-diamine

A104678-100G A104678-10G A104678-250MG A104678-25G A104678-1G A104678-5G
103796-756EA 9 USD
103796-756 103822-474 76821-466 76821-468 76821-470 76821-472 77309-336 77309-338
N,N',N''-Trimethyldiethylenetriamine 97%
N,N',N''-Trimethyldiethylenetriamine

Precaución: for Research Use Only

Formula: C₇H₁₉N₃
MW: 145.25 g/mol
Boiling Pt: 190.3±8.0 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00025637
CAS Number: 105-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR