N1,N2-Dimethylcyclohexane-1,2-diamine 96%

Supplier: Ambeed
Danger

A472944-1G A472944-25G A472944-5G A472944-10G
103828-048EA 23 USD
103828-048 103839-124 76843-038 76843-040 76843-042 76979-682
N1,N2-Dimethylcyclohexane-1,2-diamine 96%
N1,N2-Dimethylcyclohexane-1,2-diamine

Precaución: For research use only

Formula: C₈H₁₈N₂
MW: 142.2419 g/mol
Boiling Pt: 186.8 °C (760 mm Hg)
Flash Pt: 74 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05664382
CAS Number: 61798-24-1
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR