N1,N2-Dimethylbenzene-1,2-diamine 98%

Supplier: Ambeed
Warning

A386483-1G A386483-100MG A386483-250MG
103826-800EA 46 USD
103826-800 103837-842 76841-198 77168-130 77168-132
N1,N2-Dimethylbenzene-1,2-diamine 98%
N1,N2-Dimethylbenzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₈H₁₂N₂
MW: 136.19 g/mol
Melting Pt: 34.5 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD19160696
CAS Number: 3213-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR