N1,N2-Dimesitylethane-1,2-diamine 97%

Supplier: Ambeed
Warning

A129325-10G A129325-250MG A129325-5G
103801-266EA 10 USD
103801-266 76822-940 76822-942 77313-648
N1,N2-Dimesitylethane-1,2-diamine 97%
N1,N2-Dimesitylethane-1,2-diamine

Precaución: for Research Use Only

Formula: C₂₀H₂₈N₂
MW: 296.44 g/mol
Boiling Pt: 471.5 °C (760 mm Hg)
Flash Pt: 289.4 °C
Storage Temperature: Ambient
MDL Number: MFCD16877211
CAS Number: 134030-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR