N1,N1-Dimethylbenzene-1,3-diamine 97%

Supplier: Ambeed
Danger

A170426-100MG A170426-250MG A170426-5G
103806-494EA 36 USD
103806-494 103844-768 76784-206 76784-208 76784-210
N1,N1-Dimethylbenzene-1,3-diamine 97%
N1,N1-Dimethylbenzene-1,3-diamine

Precaución: for Research Use Only

Formula: C₈H₁₂N₂
Boiling Pt: 244.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00040473
CAS Number: 2836-04-6
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR