N1,N1-Dimethylbenzene-1,2-diamine 97%

Supplier: Ambeed
Danger

A277296-10G A277296-250MG A277296-1G A277296-25G A277296-5G
103799-556EA 15 USD
103799-556 103813-348 103853-282 76977-304 76977-306 77093-180 77093-182 77093-184
N1,N1-Dimethylbenzene-1,2-diamine 97%
N1,N1-Dimethylbenzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₈H₁₂N₂
Boiling Pt: 229.9 °C (760 mm Hg)
Flash Pt: 82.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01706706
CAS Number: 2836-03-5
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR