N,N-Diethyl-p-phenylenediammonium sulfate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
N,N-Diethyl-p-phenylenediamine sulphate, N,N-Diethyl-1,4-phenylenediammonium sulphate, 4-Amino-N,N-diethylaniline sulphate, N1,N1-Diethylbenzene-1,4-diamine sulphate, N,N-Diethylbenzene-1,4-diammonium sulphate
A100720-100G
A100720-10G
A100720-25G
A100720-500G
A100720-5G
103807-768EA
228
USD
103807-768
76809-208
76809-210
76809-212
76809-214
76809-216
N,N-Diethyl-p-phenylenediammonium sulfate 97%
N,N-Diethyl-p-phenylenediammonium sulphate
Precaución: for Research Use Only
Formula:
C₁₀H₁₈N₂O₄S MW: 262.33 g/mol Boiling Pt: 471.8 °C (760 mm Hg) Melting Pt: 184…186 °C Flash Pt: 239.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012993 CAS Number: 6283-63-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...