1,8-Bis(dimethylamino)naphthalene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N1,N1,N8,N8-Tetramethylnaphthalene-1,8-diamine
A137243-100G
A137243-10G
A137243-500G
A137243-5G
A137243-25G
77190-242EA
95.79
USD
77190-242
77190-244
77190-246
77298-902
77315-386
1,8-Bis(dimethylamino)naphthalene 98%
1,8-Bis(dimethylamino)naphthalene
Precaución: for Research Use Only
Formula:
C₁₄H₁₈N₂ MW: 214.31 g/mol Boiling Pt: 304.4 °C (760 mm Hg) Melting Pt: 47…51 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003920 CAS Number: 20734-58-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...