N,N,N',N'-Tetrakis(2-hydroxyethyl)adipamide 97%

Supplier: Ambeed
Warning

Synonyms: N,N,N',N'-Tetrakis(2-hydroxyethyl)hexanediamide, N1,N1,N6,N6-Tetrakis(2-hydroxyethyl)adipamide

A175988-10G A175988-25G A175988-100G
103845-420EA 5 USD
103845-420 76813-370 77082-768 77567-602
N,N,N',N'-Tetrakis(2-hydroxyethyl)adipamide 97%
N,N,N',N'-Tetrakis(2-hydroxyethyl)adipamide

Precaución: For research use only.

Formula: C₁₄H₂₈N₂O₆
MW: 320.39 g/mol
Boiling Pt: 607.7 °C (760 mm Hg)
Melting Pt: 126…130 °C
Flash Pt: 321.3 °C
Storage Temperature: Ambient
MDL Number: MFCD07787618
CAS Number: 6334-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR