N1,5-Dimethylbenzene-1,2-diamine 97%

Supplier: Ambeed
Warning

A305051-100MG A305051-250MG A305051-5G A305051-1G
103814-424EA 310 USD
103814-424 76988-854 76988-856 76988-858 77305-904
N1,5-Dimethylbenzene-1,2-diamine 97%
N1,5-Dimethylbenzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₈H₁₂N₂
MW: 136.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19203441
CAS Number: 131019-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR