EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide) 98%

Supplier: Ambeed
Danger

Synonyms: EDAC, WSC, EDC, N'-(Ethylcarbonimidoyl)-N,N-dimethylpropane-1,3-diamine, 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, N1-((Ethylimino)methylene)-N3,N3-dimethylpropane-1,3-diamine

A371088-10G A371088-25G A371088-5G
103826-472EA 5 USD
103826-472 103837-492 76773-466 76840-738 76840-740
EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide) 98%
EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide)

Precaución: for Research Use Only

Formula: C₈H₁₇N₃
MW: 155.24 g/mol
Boiling Pt: 197.7 °C (760 mm Hg)
Flash Pt: 73.4 °C
Storage Temperature: Freezer
MDL Number: MFCD00044916
CAS Number: 1892-57-5
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR