Bis(2,6-diisopropylphenyl)carbodiimide 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
N,N'-Methanediylidenebis(2,6-diisopropylaniline)
A444264-100G
A444264-10G
A444264-500G
A444264-25G
A444264-5G
103838-776EA
5
USD
103838-776
76695-176
76695-178
76695-180
76842-556
76842-558
Bis(2,6-diisopropylphenyl)carbodiimide 97%
Bis(2,6-diisopropylphenyl)carbodiimide
Precaución: for Research Use Only
Formula:
C₂₅H₃₄N₂ MW: 362.56 g/mol Boiling Pt: 477.7 °C (760 mm Hg) Melting Pt: 49…51 °C Flash Pt: 235.6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00082211 CAS Number: 2162-74-5 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...