N,N-Dimethylpyridin-4-amine 4-methylbenzenesulfonate 95%

Supplier: Ambeed
Warning

Synonyms: N,N-Dimethylpyridin-4-amine 4-methylbenzenesulphonate

A107572-100MG A107572-250MG A107572-25G A107572-1G A107572-5G
103797-216EA 14 USD
103797-216 103822-982 76821-630 76821-632 76821-634 77309-842 77309-844
N,N-Dimethylpyridin-4-amine 4-methylbenzenesulfonate 95%
N,N-Dimethylpyridin-4-amine 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₂O₃S
MW: 294.37 g/mol
Melting Pt: 176…180 °C
Storage Temperature: Ambient
MDL Number: MFCD25372259
CAS Number: 91944-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR