4-(Dimethylamino)piperidine 97%

Supplier: Ambeed
Danger

Synonyms: N,N-Dimethylpiperidin-4-amine

A836443-25G A836443-5G A836443-100G A836443-10G A836443-1G
103841-190EA 24 USD
103841-190 76845-426 76845-428 76981-768 76981-770 77308-432
4-(Dimethylamino)piperidine 97%
4-(Dimethylamino)piperidine

Precaución: For research use only

Formula: C₇H₁₆N₂
MW: 128.219 g/mol
Boiling Pt: 179.7 °C (760 mm Hg)
Flash Pt: 63.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00023144
CAS Number: 50533-97-6
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR