N,N-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine 95%

Supplier: Ambeed
Warning

Synonyms: 6-(Dimethylamino)pyridine-3-boronic acid pinacol ester

A268925-100MG A268925-250MG A268925-5G A268925-1G
103812-944EA 55 USD
103812-944 103852-848 76875-018 76875-020 77092-600 77305-028
N,N-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine 95%
N,N-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine

Precaución: for Research Use Only

Formula: C₁₃H₂₁BN₂O₂
MW: 248.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11975406
CAS Number: 1036991-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR