4-(N,N-Dimethylamino)phenylboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 4-(Dimethylamino)phenylboronic acid pinacol ester, N,N-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

A445853-1G A445853-5G A445853-10G A445853-250MG
103827-730EA 20 USD
103827-730 103838-794 76842-584 76842-586 77291-642 77291-644
4-(N,N-Dimethylamino)phenylboronic acid pinacol ester 95%
4-(N,N-Dimethylamino)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₂BNO₂
Boiling Pt: 347.1 °C (760 mm Hg)
Melting Pt: 122…123 °C
Flash Pt: 163.7 °C
Storage Temperature: Ambient
MDL Number: MFCD05663854
CAS Number: 171364-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR