N,N-Dimethyl-1H-pyrazole-1-sulfonamide 97%

Supplier: Ambeed
Warning

Synonyms: N,N-Dimethyl-1H-pyrazole-1-sulfonamide

A177821-10G A177821-250MG A177821-1G A177821-5G A177821-25G
103807-270EA 19 USD
103807-270 103845-604 76813-438 76813-440 77082-994 77082-996 77671-520
N,N-Dimethyl-1H-pyrazole-1-sulfonamide 97%
N,N-Dimethyl-1H-pyrazole-1-sulfonamide

Precaución: for Research Use Only

Formula: C₅H₉N₃O₂S
MW: 175.21 g/mol
Boiling Pt: 287.4 °C (760 mm Hg)
Flash Pt: 127.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11865226
CAS Number: 133228-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR