N,N-Dimethyl-1-(5-methyl-1H-indol-3-yl)methanamine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Methylgramine

A100183-250MG A100183-1G
103795-284EA 75 USD
103795-284 103820-996 76765-096 77296-586
N,N-Dimethyl-1-(5-methyl-1H-indol-3-yl)methanamine 97%
N,N-Dimethyl-1-(5-methyl-1H-indol-3-yl)methanamine

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂
MW: 188.27 g/mol
Boiling Pt: 311.5 °C (760 mm Hg)
Flash Pt: 142.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00022732
CAS Number: 30218-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR