1,3-Bis(tert-butoxycarbonyl)thiourea 96%

Supplier: Ambeed
Warning

Synonyms: 1,3-Di-Boc-thiourea

A116610-100G A116610-100MG A116610-250MG A116610-25G A116610-1G A116610-5G A116610-10G
103798-766EA 8 USD
103798-766 103824-630 76718-322 76718-324 76718-326 76718-328 77297-478 77297-480 77571-006
1,3-Bis(tert-butoxycarbonyl)thiourea 96%
1,3-Bis(tert-butoxycarbonyl)thiourea

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₄S
MW: 276.35 g/mol
Melting Pt: 131…135 °C
Storage Temperature: Ambient
MDL Number: MFCD03093917
CAS Number: 145013-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR